This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288184_gH
See other HYI GE Assays ›
Gene Symbol:
HYI
Gene Name:
hydroxypyruvate isomerase (putative)
Gene Aliases:
HT036
Chromosome Location:
Chr.1: 43450607 - 43454241 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288184_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HYI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190880.2 NP_001177809.1 4 - 5 702 87
NM_001243526.1 NP_001230455.1 5 - 6 777 87
NM_031207.5 NP_112484.3 4 - 5 702 87
XM_005271239.4 XP_005271296.1 5 - 6 1029 87
XM_005271240.4 XP_005271297.1 4 - 5 927 87
XM_006710937.3 XP_006711000.1 5 - 6 1029 87
XM_011542222.2 XP_011540524.1 4 - 5 927 87
GenBank mRNA AF284750.1 - 4 - 5 577 87
AF284751.1 - 4 - 5 577 87
AK096244.1 - 2 - 3 1778 87
AY037165.1 - 4 - 5 676 87
AY775560.1 - 4 - 5 677 87
BC006140.1 - 4 - 5 388 3928690 87
BC019041.2 - 4 - 5 504 4586546 87
BM826556.1 - 5 - 6 444 87
CD672533.1 - 4 - 5 430 87
JF911821.1 - 3 - 4 287 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HYI

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding
hydroxypyruvate isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline