This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288437_g1
See other GTF2H4 GE Assays ›
Gene Symbol:
GTF2H4
Gene Name:
general transcription factor IIH subunit 4
Gene Aliases:
P52, TFB2, TFIIH
Chromosome Location:
Chr.6: 30908200 - 30914103 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288437_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2H4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001517.4 NP_001508.1 10 - 11 1170 89
GenBank mRNA AK222607.1 - 10 - 11 1074 89
AK298058.1 - 10 - 11 1218 89
BC004935.1 - 10 - 11 1142 3606718 89
BC016302.2 - 10 - 11 1142 3830902 89
BT007321.1 - 9 - 10 963 89
Y07595.1 - 10 - 11 1090 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2H4

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
nucleotide-excision repair, DNA incision
phosphorylation of RNA polymerase II C-terminal domain
global genome nucleotide-excision repair
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
ATP-dependent DNA helicase activity
protein kinase activity
protein binding
DNA-dependent ATPase activity
RNA polymerase II carboxy-terminal domain kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0