This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288563_m1
See other ATP5S GE Assays ›
Gene Symbol:
ATP5S
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases:
ATPW, FB, HSU79253
Chromosome Location:
Chr.14: 50311752 - 50326715 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288563_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5S
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003805.2 NP_001003805.1 3 - 4 1055 80
XM_011536657.2 XP_011534959.1 3 - 4 493 80
GenBank mRNA BC011549.1 - 3 - 4 1055 4554461 80
BC087851.1 - 4 - 5 860 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5S

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
mitochondrial ATP synthesis coupled proton transport
hydrogen ion transmembrane transporter activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0