This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288658_g1
See other RBM4 GE Assays ›
Gene Symbol:
RBM4
Gene Name:
RNA binding motif protein 4
Gene Aliases:
LARK, RBM4A, ZCCHC21, ZCRB3A
Chromosome Location:
Chr.11: 66638617 - 66668385 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288658_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198843.1 NP_001185772.1 1 - 2 136 98
NM_001198844.1 NP_001185773.1 1 - 2 136 98
NM_002896.3 NP_002887.2 1 - 2 136 98
GenBank mRNA DA046115.1 - 1 - 2 100 98
DC366746.1 - 1 - 2 136 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001198843.1, NM_001198844.1, NM_002896.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM4

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
cap-independent translational initiation
IRES-dependent translational initiation
RNA processing
mRNA processing
RNA splicing
negative regulation of translation
cell differentiation
negative regulation of translation in response to stress
miRNA mediated inhibition of translation
entrainment of circadian clock by photoperiod
negative regulation of translational initiation
response to arsenic-containing substance
regulation of nucleocytoplasmic transport
positive regulation of muscle cell differentiation
stress-activated MAPK cascade
circadian regulation of translation
nucleotide binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
zinc ion binding
miRNA binding
poly(A) RNA binding
pre-mRNA intronic binding
pre-mRNA intronic pyrimidine-rich binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0