This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288697_m1
See other SEC22B GE Assays ›
Gene Symbol:
SEC22B
Gene Name:
SEC22 homolog B, vesicle trafficking protein (gene/pseudogene)
Gene Aliases:
ERS-24, SEC22L1
Chromosome Location:
Chr.1: 120152520 - 120176534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288697_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC22B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004892.5 NP_004883.3 1 - 2 219 63
XM_017002856.1 XP_016858345.1 1 - 2 232 63
GenBank mRNA AK289875.1 - 1 - 2 213 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SEC22B

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport
regulation of organelle organization
positive regulation of protein catabolic process
COPII vesicle coating
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0