This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288822_g1
See other NDOR1 GE Assays ›
Gene Symbol:
NDOR1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
NR1, bA350O14.9
Chromosome Location:
Chr.9: 137205667 - 137219361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144026.2 NP_001137498.1 10 - 11 1401 94
NM_001144027.2 NP_001137499.1 9 - 10 1299 94
NM_001144028.2 NP_001137500.1 10 - 11 1380 94
NM_014434.3 NP_055249.1 10 - 11 1401 94
XM_011518545.1 XP_011516847.1 9 - 10 1289 94
XM_011518547.1 XP_011516849.1 10 - 11 1191 94
XM_011518548.2 XP_011516850.1 9 - 10 2591 94
XM_017014626.1 XP_016870115.1 10 - 11 1401 94
XM_017014627.1 XP_016870116.1 10 - 11 1401 94
GenBank mRNA AF199509.1 - 10 - 11 1337 94
AK026089.1 - 10 - 11 1373 94
AK074403.1 - 9 - 10 1260 94
AK225995.1 - 10 - 11 1372 94
AK290026.1 - 10 - 11 1398 94
AY077845.1 - 10 - 11 1319 94
BC015735.1 - 10 - 11 1337 4869561 94
BC078668.1 - 10 - 11 1337 94
BC093782.1 - 10 - 11 1318 7939627 94
BC111943.1 - 10 - 11 1318 8327411 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDOR1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
iron-sulfur cluster assembly
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
protein binding
FMN binding
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0