This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288825_g1
See other NDOR1 GE Assays ›
Gene Symbol:
NDOR1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
NR1, bA350O14.9
Chromosome Location:
Chr.9: 137205667 - 137219361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288825_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144026.2 NP_001137498.1 2 - 3 327 78
NM_001144027.2 NP_001137499.1 2 - 3 327 78
NM_001144028.2 NP_001137500.1 2 - 3 327 78
NM_014434.3 NP_055249.1 2 - 3 327 78
XM_011518545.1 XP_011516847.1 2 - 3 317 78
XM_011518548.2 XP_011516850.1 2 - 3 306 78
XM_017014626.1 XP_016870115.1 2 - 3 327 78
XM_017014627.1 XP_016870116.1 2 - 3 327 78
GenBank mRNA AF199509.1 - 2 - 3 263 78
AK026089.1 - 2 - 3 299 78
AK074403.1 - 2 - 3 288 78
AK225995.1 - 2 - 3 298 78
AK290026.1 - 2 - 3 324 78
AY077845.1 - 2 - 3 245 78
BC015735.1 - 2 - 3 263 4869561 78
BC078668.1 - 2 - 3 263 78
BC093782.1 - 2 - 3 244 7939627 78
BC111943.1 - 2 - 3 244 8327411 78
DA408370.1 - 2 - 3 327 78
DA858250.1 - 2 - 3 327 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011518548.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDOR1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
iron-sulfur cluster assembly
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
protein binding
FMN binding
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0