This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288827_g1
See other NDOR1 GE Assays ›
Gene Symbol:
NDOR1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
NR1, bA350O14.9
Chromosome Location:
Chr.9: 137205667 - 137219361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288827_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144026.2 NP_001137498.1 4 - 5 520 101
NM_001144028.2 NP_001137500.1 4 - 5 520 101
NM_014434.3 NP_055249.1 4 - 5 520 101
XM_011518547.1 XP_011516849.1 4 - 5 310 101
XM_011518548.2 XP_011516850.1 3 - 4 1679 101
XM_017014626.1 XP_016870115.1 4 - 5 520 101
XM_017014627.1 XP_016870116.1 4 - 5 520 101
GenBank mRNA AF199509.1 - 4 - 5 456 101
AK026089.1 - 4 - 5 492 101
AK225995.1 - 4 - 5 491 101
AK290026.1 - 4 - 5 517 101
AY077845.1 - 4 - 5 438 101
BC015735.1 - 4 - 5 456 4869561 101
BC078668.1 - 4 - 5 456 101
BC093782.1 - 4 - 5 437 7939627 101
BC111943.1 - 4 - 5 437 8327411 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011518548.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDOR1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
iron-sulfur cluster assembly
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
protein binding
FMN binding
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0