This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288831_g1
See other NDOR1 GE Assays ›
Gene Symbol:
NDOR1
Gene Name:
NADPH dependent diflavin oxidoreductase 1
Gene Aliases:
NR1, bA350O14.9
Chromosome Location:
Chr.9: 137205667 - 137219361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288831_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDOR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144026.2 NP_001137498.1 9 - 10 1282 108
NM_001144027.2 NP_001137499.1 8 - 9 1180 108
NM_014434.3 NP_055249.1 9 - 10 1282 108
XM_011518545.1 XP_011516847.1 8 - 9 1170 108
XM_011518547.1 XP_011516849.1 9 - 10 1072 108
XM_011518548.2 XP_011516850.1 8 - 9 2472 108
XM_017014626.1 XP_016870115.1 9 - 10 1282 108
XM_017014627.1 XP_016870116.1 9 - 10 1282 108
GenBank mRNA AF199509.1 - 9 - 10 1218 108
AK026089.1 - 9 - 10 1254 108
AK074403.1 - 8 - 9 1141 108
AK225995.1 - 9 - 10 1253 108
AK290026.1 - 9 - 10 1279 108
AY077845.1 - 9 - 10 1200 108
BC015735.1 - 9 - 10 1218 4869561 108
BC078668.1 - 9 - 10 1218 108
BC093782.1 - 9 - 10 1199 7939627 108
BC111943.1 - 9 - 10 1199 8327411 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDOR1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

cell death
iron-sulfur cluster assembly
cellular response to menadione
oxidation-reduction process
NADPH-hemoprotein reductase activity
protein binding
FMN binding
oxidoreductase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0