This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288875_g1
See other METTL26 GE Assays ›
Gene Symbol:
METTL26
Gene Name:
methyltransferase like 26
Gene Aliases:
C16orf13, JFP2
Chromosome Location:
Chr.16: 634427 - 636366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288875_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040160.2 NP_001035250.1 4 - 5 575 64
NM_001040161.2 NP_001035251.1 2 - 3 352 64
NM_001040162.2 NP_001035252.1 3 - 4 412 64
NM_001040165.2 NP_001035255.1 3 - 4 515 64
NM_001288710.1 NP_001275639.1 4 - 5 583 64
NM_032366.4 NP_115742.3 5 - 6 643 64
NR_109976.1 4 - 5 587 64
NR_109977.1 4 - 5 480 64
NR_109978.1 5 - 6 750 64
NR_109979.1 3 - 4 420 64
XM_011522713.2 XP_011521015.1 5 - 6 842 64
XM_011522714.2 XP_011521016.1 4 - 5 667 64
GenBank mRNA AI146806.1 - 3 - 4 409 64
AK057077.1 - 2 - 3 1541 64
AK294482.1 - 5 - 6 579 64
AK295628.1 - 4 - 5 511 64
BC007207.1 - 2 - 3 334 4054799 64
BI762987.1 - 5 - 6 619 64
BM827705.1 - 3 - 4 347 64
BP276058.1 - 4 - 5 542 64
BQ921757.1 - 548 64
BU539308.1 - 3 - 4 368 64
BX104467.1 - 1 - 2 231 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL26

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0