This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288876_g1
See other METTL26 GE Assays ›
Gene Symbol:
METTL26
Gene Name:
methyltransferase like 26
Gene Aliases:
C16orf13, JFP2
Chromosome Location:
Chr.16: 634427 - 636366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040160.2 NP_001035250.1 1 - 2 267 103
NM_001040165.2 NP_001035255.1 1 - 2 267 103
NM_001288710.1 NP_001275639.1 1 - 2 267 103
NM_032366.4 NP_115742.3 1 - 2 267 103
NR_109978.1 1 - 2 267 103
XM_011522713.2 XP_011521015.1 1 - 2 265 103
XM_011522714.2 XP_011521016.1 1 - 2 265 103
GenBank mRNA AI146806.1 - 1 - 2 161 103
AK294482.1 - 1 - 2 203 103
AK295628.1 - 1 - 2 203 103
BI762987.1 - 1 - 2 137 103
BP273946.1 - 1 - 2 203 103
BP276058.1 - 1 - 2 234 103
BQ921757.1 - 172 103
CK904040.1 - 1 - 2 171 103
DB499720.1 - 1 - 2 267 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL26

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0