This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288888_m1
See other PTPN20 GE Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288888_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042362.3 NP_001035821.1 4 - 5 425 73
NM_001042365.3 NP_001035824.1 2 - 3 217 73
NM_001320682.1 NP_001307611.1 3 - 4 315 73
NM_001320689.1 NP_001307618.1 4 - 5 466 73
NM_001320690.1 NP_001307619.1 3 - 4 330 73
XM_017016060.1 XP_016871549.1 5 - 6 546 73
XM_017016061.1 XP_016871550.1 4 - 5 496 73
XM_017016062.1 XP_016871551.1 4 - 5 616 73
XM_017016063.1 XP_016871552.1 5 - 6 587 73
XM_017016064.1 XP_016871553.1 3 - 4 339 73
XM_017016065.1 XP_016871554.1 4 - 5 389 73
GenBank mRNA AY704145.1 - 4 - 5 366 73
AY704149.1 - 4 - 5 415 73
AY704155.1 - 4 - 5 456 73
AY704156.1 - 4 - 5 407 73
AY704157.1 - 3 - 4 271 73
BC143394.1 - 4 - 5 421 73
BC171725.1 - 4 - 5 421 73
BX648913.1 - 2 - 3 218 73
CR933668.1 - 3 - 4 283 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0