This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288896_m1
See other SMIM8 GE Assays ›
Gene Symbol:
SMIM8
Gene Name:
small integral membrane protein 8
Gene Aliases:
C6orf162, dJ102H19.2
Chromosome Location:
Chr.6: 87322588 - 87342328 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288896_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMIM8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042493.2 NP_001035958.1 2 - 3 58 73
GenBank mRNA AK311963.1 - 2 - 3 29 73
DA293899.1 - 2 - 3 29 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK311963.1, NM_001042493.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0