Assay Details
| Gene Symbol: FGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173558.3 | NP_775829.2 | 10 - 11 | 1372 | 103 | |
| XM_017010429.1 | XP_016865918.1 | 10 - 11 | 1364 | 103 | ||
| GenBank mRNA | AK092732.1 | - | 9 - 10 | 1581 | 103 | |
| AK097981.1 | - | 6 - 7 | 1256 | 103 | ||
| AK131079.1 | - | 5 - 6 | 1200 | 103 | ||
| AK290419.1 | - | 11 - 12 | 1365 | 103 | ||
| AK301156.1 | - | 8 - 9 | 1105 | 103 | ||
| BC023645.2 | - | 10 - 11 | 1372 | 5087501 | 103 | |
| BC053655.1 | - | 11 - 12 | 1317 | 5755893 | 103 | |
| BX648164.1 | - | 10 - 11 | 1279 | 103 | ||
More Information
Set Membership: |
|

FGD2
Genomic Map