This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289681_m1
See other DCAF4 GE Assays ›
Gene Symbol:
DCAF4
Gene Name:
DDB1 and CUL4 associated factor 4
Gene Aliases:
WDR21, WDR21A
Chromosome Location:
Chr.14: 72926332 - 72962098 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289681_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCAF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163508.1 NP_001156980.1 9 - 10 1028 71
NM_001163509.1 NP_001156981.1 9 - 10 996 71
NM_181341.2 NP_851938.2 7 - 8 845 71
XM_017021201.1 XP_016876690.1 8 - 9 837 71
XM_017021202.1 XP_016876691.1 8 - 9 837 71
XM_017021203.1 XP_016876692.1 9 - 10 1023 71
XM_017021204.1 XP_016876693.1 9 - 10 993 71
XM_017021205.1 XP_016876694.1 10 - 11 1162 71
XM_017021206.1 XP_016876695.1 9 - 10 998 71
XM_017021209.1 XP_016876698.1 9 - 10 1023 71
XM_017021211.1 XP_016876700.1 9 - 10 967 71
XM_017021212.1 XP_016876701.1 10 - 11 1130 71
XM_017021213.1 XP_016876702.1 9 - 10 930 71
GenBank mRNA AK027745.1 - 9 - 10 873 71
AK297571.1 - 9 - 10 873 71
AK297742.1 - 9 - 10 839 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DCAF4

Gene Ontology Categories:

Function(s)

protein ubiquitination

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xxb7z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline