This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289694_g1
See other EIF6 GE Assays ›
Gene Symbol:
EIF6
Gene Name:
eukaryotic translation initiation factor 6
Gene Aliases:
CAB, EIF3A, ITGB4BP, b(2)gcn, eIF-6, p27(BBP), p27BBP
Chromosome Location:
Chr.20: 35278906 - 35284816 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289694_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267810.1 NP_001254739.1 2 - 3 128 117
NM_002212.3 NP_002203.1 2 - 3 198 117
NM_181468.2 NP_852133.1 1 - 2 332 117
NR_052022.1 2 - 3 198 117
NR_052023.1 2 - 3 198 117
GenBank mRNA AB062289.1 - 2 - 3 131 117
AF022229.1 - 1 - 2 187 117
AF047433.1 - 2 - 3 172 117
AK314969.1 - 2 - 3 174 117
BC001119.2 - 2 - 3 171 2989973 117
BC011845.2 - 2 - 3 149 4120463 117
BC019305.1 - 2 - 3 131 2823211 117
BC095465.1 - 2 - 3 147 117
BQ222221.1 - 1 - 2 118 117
CN265282.1 - 1 - 2 332 117
CR456764.1 - 1 - 2 102 117
CT002623.1 - 2 - 3 150 117
DB448068.1 - 2 - 3 199 117
Y11435.1 - 2 - 3 140 117

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF6

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal subunit export from nucleus
maturation of 5.8S rRNA
maturation of LSU-rRNA
regulation of glycolytic process
translational initiation
response to insulin
gene silencing by miRNA
miRNA mediated inhibition of translation
mature ribosome assembly
regulation of fatty acid biosynthetic process
regulation of megakaryocyte differentiation
positive regulation of translation
assembly of large subunit precursor of preribosome
regulation of reactive oxygen species metabolic process
translation initiation factor activity
protein binding
ribosome binding
ribosomal large subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0