This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289732_m1
See other GPX6 GE Assays ›
Gene Symbol:
GPX6
Gene Name:
glutathione peroxidase 6
Gene Aliases:
GPX5p, GPXP3, GPx-6, GSHPx-6, dJ1186N24, dJ1186N24.1
Chromosome Location:
Chr.6: 28503296 - 28515793 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289732_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182701.1 NP_874360.1 3 - 4 415 96
GenBank mRNA AY324826.1 - 3 - 4 415 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPX6

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
oxidation-reduction process
cellular oxidant detoxification
glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-drmdc:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0