This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289752_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289752_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 10 - 11 1427 92
NM_001199506.1 NP_001186435.1 10 - 11 1422 92
NM_001281507.1 NP_001268436.1 10 - 11 1526 92
NM_080672.4 NP_542403.1 10 - 11 1904 92
NM_183244.1 NP_899067.1 10 - 11 1547 92
NM_183246.1 NP_899069.1 9 - 10 1337 92
XM_011528525.2 XP_011526827.1 10 - 11 1533 92
XM_011528526.2 XP_011526828.1 9 - 10 1339 92
XM_017027626.1 XP_016883115.1 10 - 11 1636 92
XM_017027627.1 XP_016883116.1 10 - 11 1581 92
XM_017027628.1 XP_016883117.1 9 - 10 1229 92
XM_017027629.1 XP_016883118.1 9 - 10 1372 92
XM_017027630.1 XP_016883119.1 9 - 10 1200 92
XM_017027631.1 XP_016883120.1 10 - 11 1463 92
GenBank mRNA AB098521.1 - 10 - 11 1547 92
AB098522.1 - 9 - 10 1337 92
AJ311122.2 - 10 - 11 1903 92
AJ617581.1 - 10 - 11 1422 92
AK091632.1 - 10 - 11 1549 92
AK098788.1 - 10 - 11 1427 92
AK295459.1 - 10 - 11 1526 92
AK314493.1 - 10 - 11 1634 92
AK316047.1 - 10 - 11 1419 92
BC108303.1 - 10 - 11 1904 4179728 92
BC117362.1 - 10 - 11 1792 40125913 92
BC117364.1 - 10 - 11 1795 40125915 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0