This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289753_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289753_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183244.1 NP_899067.1 1 - 2 227 95
NM_183246.1 NP_899069.1 1 - 2 227 95
GenBank mRNA AB098521.1 - 1 - 2 227 95
AB098522.1 - 1 - 2 227 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB098521.1, AB098522.1, NM_183244.1, NM_183246.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0