This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289754_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289754_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 3 - 4 348 117
NM_001199506.1 NP_001186435.1 3 - 4 343 117
NM_001281507.1 NP_001268436.1 3 - 4 447 117
NM_080672.4 NP_542403.1 3 - 4 825 117
NM_183244.1 NP_899067.1 3 - 4 468 117
NM_183246.1 NP_899069.1 3 - 4 468 117
XM_011528525.2 XP_011526827.1 3 - 4 454 117
XM_011528526.2 XP_011526828.1 3 - 4 470 117
XM_017027626.1 XP_016883115.1 3 - 4 557 117
XM_017027627.1 XP_016883116.1 3 - 4 502 117
XM_017027628.1 XP_016883117.1 3 - 4 360 117
XM_017027629.1 XP_016883118.1 3 - 4 503 117
XM_017027630.1 XP_016883119.1 3 - 4 331 117
XM_017027631.1 XP_016883120.1 3 - 4 384 117
GenBank mRNA AB098521.1 - 3 - 4 468 117
AB098522.1 - 3 - 4 468 117
AJ311122.2 - 3 - 4 824 117
AJ617581.1 - 3 - 4 343 117
AK091632.1 - 3 - 4 470 117
AK295459.1 - 3 - 4 447 117
AK314493.1 - 3 - 4 555 117
AK316047.1 - 3 - 4 340 117
BC108303.1 - 3 - 4 825 4179728 117
BC117362.1 - 3 - 4 713 40125913 117
BC117364.1 - 3 - 4 716 40125915 117

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0