This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289755_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289755_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 5 - 6 741 61
NM_001199506.1 NP_001186435.1 5 - 6 736 61
NM_001281507.1 NP_001268436.1 5 - 6 840 61
NM_080672.4 NP_542403.1 5 - 6 1218 61
NM_183244.1 NP_899067.1 5 - 6 861 61
XM_011528525.2 XP_011526827.1 5 - 6 847 61
XM_017027626.1 XP_016883115.1 5 - 6 950 61
XM_017027627.1 XP_016883116.1 5 - 6 895 61
XM_017027631.1 XP_016883120.1 5 - 6 777 61
GenBank mRNA AB098521.1 - 5 - 6 861 61
AJ311122.2 - 5 - 6 1217 61
AJ617581.1 - 5 - 6 736 61
AK091632.1 - 5 - 6 863 61
AK098788.1 - 5 - 6 741 61
AK295459.1 - 5 - 6 840 61
AK314493.1 - 5 - 6 948 61
AK316047.1 - 5 - 6 733 61
BC108303.1 - 5 - 6 1218 4179728 61
BC117362.1 - 5 - 6 1106 40125913 61
BC117364.1 - 5 - 6 1109 40125915 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x4xlz:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0