This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289756_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289756_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 6 - 7 917 116
NM_001199506.1 NP_001186435.1 6 - 7 912 116
NM_001281507.1 NP_001268436.1 6 - 7 1016 116
NM_080672.4 NP_542403.1 6 - 7 1394 116
NM_183244.1 NP_899067.1 6 - 7 1037 116
NM_183246.1 NP_899069.1 5 - 6 827 116
XM_011528525.2 XP_011526827.1 6 - 7 1023 116
XM_011528526.2 XP_011526828.1 5 - 6 829 116
XM_017027626.1 XP_016883115.1 6 - 7 1126 116
XM_017027627.1 XP_016883116.1 6 - 7 1071 116
XM_017027628.1 XP_016883117.1 5 - 6 719 116
XM_017027629.1 XP_016883118.1 5 - 6 862 116
XM_017027630.1 XP_016883119.1 5 - 6 690 116
XM_017027631.1 XP_016883120.1 6 - 7 953 116
GenBank mRNA AJ311122.2 - 6 - 7 1393 116
AK091632.1 - 6 - 7 1039 116
AK098788.1 - 6 - 7 917 116
AK295459.1 - 6 - 7 1016 116
AK314493.1 - 6 - 7 1124 116
AK316047.1 - 6 - 7 909 116
BC108303.1 - 6 - 7 1394 4179728 116
BC117362.1 - 6 - 7 1282 40125913 116
BC117364.1 - 6 - 7 1285 40125915 116

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0