This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289777_m1
See other ALDOA GE Assays ›
Gene Symbol:
ALDOA
Gene Name:
aldolase, fructose-bisphosphate A
Gene Aliases:
ALDA, GSD12, HEL-S-87p
Chromosome Location:
Chr.16: 30053090 - 30070420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289777_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_184041.2 NP_908930.1 1 - 2 258 108
GenBank mRNA AK026577.1 - 1 - 2 108 108
BC010660.1 - 1 - 2 176 3869603 108
BC012880.1 - 1 - 2 104 3459009 108
BC015888.2 - 1 - 2 100 3911623 108
BQ719865.1 - 1 - 2 219 108
D28356.1 - 1 - 2 169 108
DA138954.1 - 1 - 2 258 108
X05236.1 - 1 - 2 128 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC010660.1, BC012880.1, BC015888.2, D28356.1, NM_184041.2, X05236.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALDOA

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

platelet degranulation
fructose metabolic process
gluconeogenesis
glycolytic process
ATP biosynthetic process
striated muscle contraction
actin filament organization
regulation of cell shape
fructose 1,6-bisphosphate metabolic process
muscle cell cellular homeostasis
protein homotetramerization
canonical glycolysis
cell-cell adhesion
actin binding
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding
tubulin binding
identical protein binding
poly(A) RNA binding
fructose binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0