This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289823_m1
See other NUDT1 GE Assays ›
Gene Symbol:
NUDT1
Gene Name:
nudix hydrolase 1
Gene Aliases:
MTH1
Chromosome Location:
Chr.7: 2242222 - 2251145 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289823_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198949.1 NP_945187.1 2 - 3 165 80
NM_198952.1 NP_945190.1 2 - 3 162 80
NM_198954.1 NP_945192.1 1 - 2 150 80
GenBank mRNA AB025235.1 - 2 - 3 146 80
AB025237.1 - 2 - 3 162 80
AB025239.1 - 1 - 2 150 80
AB025240.1 - 2 - 3 146 80
AB025241.1 - 2 - 3 162 80
AB025242.1 - 1 - 2 150 80
BC014618.2 - 1 - 2 144 3531786 80
BC065367.1 - 1 - 2 249 5814797 80
BM927392.1 - 1 - 2 248 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUDT1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide catabolic process
dGTP catabolic process
DNA repair
response to oxidative stress
nucleobase-containing small molecule catabolic process
DNA protection
dATP catabolic process
GTPase activity
protein binding
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
snoRNA binding
8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
GTP diphosphatase activity
2-hydroxy-adenosine triphosphate pyrophosphatase activity
2-hydroxy-(deoxy)adenosine-triphosphate pyrophosphatase activity
metal ion binding
ATP diphosphatase activity
m7G(5')pppN diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0