This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289825_m1
See other NUDT1 GE Assays ›
Gene Symbol:
NUDT1
Gene Name:
nudix hydrolase 1
Gene Aliases:
MTH1
Chromosome Location:
Chr.7: 2242222 - 2251145 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289825_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198950.1 NP_945188.1 2 - 3 89 108
GenBank mRNA AB025236.1 - 2 - 3 89 108
AK026631.1 - 2 - 3 91 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB025236.1, NM_198950.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUDT1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide catabolic process
dGTP catabolic process
DNA repair
response to oxidative stress
nucleobase-containing small molecule catabolic process
DNA protection
dATP catabolic process
GTPase activity
protein binding
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
snoRNA binding
8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
GTP diphosphatase activity
2-hydroxy-adenosine triphosphate pyrophosphatase activity
2-hydroxy-(deoxy)adenosine-triphosphate pyrophosphatase activity
metal ion binding
ATP diphosphatase activity
m7G(5')pppN diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0