This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289908_m1
See other TMIGD1 GE Assays ›
Gene Symbol:
TMIGD1
Gene Name:
transmembrane and immunoglobulin domain containing 1
Gene Aliases:
TMIGD, UNQ9372
Chromosome Location:
Chr.17: 30316329 - 30334076 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289908_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMIGD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_206832.2 NP_996663.1 4 - 5 732 75
XM_011524787.1 XP_011523089.1 4 - 5 746 75
GenBank mRNA AK290266.1 - 4 - 5 705 75
AY358153.1 - 4 - 5 720 75
BC137201.1 - 4 - 5 716 9021198 75
BC137202.1 - 4 - 5 716 9021199 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMIGD1

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0