This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290125_m1
See other CYBRD1 GE Assays ›
Gene Symbol:
CYBRD1
Gene Name:
cytochrome b reductase 1
Gene Aliases:
CYB561A2, DCYTB, FRRS3
Chromosome Location:
Chr.2: 171522247 - 171558133 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290125_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYBRD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127383.1 NP_001120855.1 1 - 2 383 67
GenBank mRNA AK128495.1 - 1 - 2 262 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

cellular iron ion homeostasis
response to iron ion
oxidation-reduction process
ferric-chelate reductase activity
protein binding
oxidoreductase activity, oxidizing metal ions
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline