This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290129_m1
See other ALDOA GE Assays ›
Gene Symbol:
ALDOA
Gene Name:
aldolase, fructose-bisphosphate A
Gene Aliases:
ALDA, GSD12, HEL-S-87p
Chromosome Location:
Chr.16: 30053090 - 30070420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290129_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127617.2 NP_001121089.1 1 - 2 285 57
GenBank mRNA BX647566.1 - 1 - 2 74 57
DA893541.1 - 1 - 2 132 57
DA899482.1 - 1 - 2 285 57

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001127617.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

platelet degranulation
fructose metabolic process
gluconeogenesis
glycolytic process
ATP biosynthetic process
striated muscle contraction
actin filament organization
regulation of cell shape
fructose 1,6-bisphosphate metabolic process
muscle cell cellular homeostasis
protein homotetramerization
canonical glycolysis
cell-cell adhesion
actin binding
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding
tubulin binding
identical protein binding
poly(A) RNA binding
fructose binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0