This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290155_m1
See other PPP1R14D GE Assays ›
Gene Symbol:
PPP1R14D
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14D
Gene Aliases:
CPI17-like, GBPI-1, GBPI1
Chromosome Location:
Chr.15: 40815445 - 40828793 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290155_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130143.1 NP_001123615.1 1 - 2 322 59
GenBank mRNA BC098150.1 - 1 - 2 328 59
BC098341.1 - 1 - 2 328 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC098150.1, BC098341.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
regulation of phosphorylation
negative regulation of catalytic activity
protein serine/threonine phosphatase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0