This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290426_m1
See other COX11 GE Assays ›
Gene Symbol:
COX11
Gene Name:
COX11, cytochrome c oxidase copper chaperone
Gene Aliases:
COX11P
Chromosome Location:
Chr.17: 54951898 - 54971024 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290426_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162861.2 NP_001156333.1 3 - 4 791 100
NR_027941.2 3 - 4 791 100
GenBank mRNA BM975167.1 - 2 - 3 431 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

respiratory gaseous exchange
respiratory chain complex IV assembly
aerobic respiration
negative regulation of glucokinase activity
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
copper ion binding
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0