This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290473_g1
See other HDAC8 GE Assays ›
Gene Symbol:
HDAC8
Gene Name:
histone deacetylase 8
Gene Aliases:
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
Chromosome Location:
Chr.X: 72329516 - 72573103 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290473_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166418.1 NP_001159890.1 1 - 2 455 89
NM_001166419.1 NP_001159891.1 1 - 2 455 89
NM_001166420.1 NP_001159892.1 1 - 2 455 89
NM_001166422.1 NP_001159894.1 1 - 2 455 89
NM_001166448.1 NP_001159920.1 1 - 2 455 89
NM_018486.2 NP_060956.1 1 - 2 455 89
NR_051952.1 1 - 2 455 89
XM_011530986.2 XP_011529288.2 1 - 2 450 89
XM_011530987.2 XP_011529289.2 1 - 2 450 89
XM_017029640.1 XP_016885129.1 1 - 2 450 89
XM_017029641.1 XP_016885130.1 1 - 2 450 89
XM_017029642.1 XP_016885131.1 1 - 2 448 89
XM_017029643.1 XP_016885132.1 1 - 2 450 89
XM_017029644.1 XP_016885133.1 1 - 2 450 89
XM_017029645.1 XP_016885134.1 1 - 2 450 89
XM_017029646.1 XP_016885135.1 1 - 2 130 89
XM_017029647.1 XP_016885136.1 1 - 2 450 89
GenBank mRNA AF212246.1 - 1 - 2 171 89
AF230097.1 - 1 - 2 168 89
AF245664.1 - 1 - 2 122 89
AJ277724.1 - 1 - 2 155 89
AK074326.1 - 1 - 2 400 89
AK294903.1 - 1 - 2 169 89
AK296641.1 - 1 - 2 405 89
AK298767.1 - 1 - 2 195 89
AK300895.1 - 1 - 2 152 89
AY429539.1 - 1 - 2 130 89
BC050433.1 - 1 - 2 146 5761745 89
BP229332.1 - 1 - 2 431 89
BQ189619.1 - 1 - 2 205 89
BQ632790.1 - 1 - 2 167 89
DC352390.1 - 1 - 2 456 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK298767.1, XM_017029646.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDAC8

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin assembly or disassembly
transcription, DNA-templated
sister chromatid cohesion
chromatin modification
negative regulation of protein ubiquitination
regulation of protein stability
regulation of telomere maintenance
histone H3 deacetylation
regulation of cohesin loading
histone deacetylase activity
protein binding
transcription factor binding
Hsp70 protein binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
metal ion binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline