This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290474_g1
See other HDAC8 GE Assays ›
Gene Symbol:
HDAC8
Gene Name:
histone deacetylase 8
Gene Aliases:
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
Chromosome Location:
Chr.X: 72329516 - 72573103 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290474_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166418.1 NP_001159890.1 2 - 3 508 97
NM_001166448.1 NP_001159920.1 2 - 3 508 97
XM_017029642.1 XP_016885131.1 2 - 3 501 97
XM_017029644.1 XP_016885133.1 2 - 3 503 97
GenBank mRNA AK294903.1 - 2 - 3 222 97
AK296641.1 - 2 - 3 458 97
BQ632790.1 - 2 - 3 220 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chromatin assembly or disassembly
transcription, DNA-templated
sister chromatid cohesion
chromatin modification
negative regulation of protein ubiquitination
regulation of protein stability
regulation of telomere maintenance
histone H3 deacetylation
regulation of cohesin loading
histone deacetylase activity
protein binding
transcription factor binding
Hsp70 protein binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
metal ion binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline