This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290511_m1
See other MYD88 GE Assays ›
Gene Symbol:
MYD88
Gene Name:
myeloid differentiation primary response 88
Gene Aliases:
MYD88D
Chromosome Location:
Chr.3: 38138478 - 38143022 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290511_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYD88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172566.1 NP_001166037.1 1 - 2 551 96
GenBank mRNA AK298650.1 - 1 - 2 372 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

toll-like receptor signaling pathway
MyD88-dependent toll-like receptor signaling pathway
apoptotic process
inflammatory response
signal transduction
cell surface receptor signaling pathway
positive regulation of type I interferon production
positive regulation of interleukin-17 production
positive regulation of interleukin-23 production
positive regulation of interleukin-6 production
toll-like receptor 9 signaling pathway
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
regulation of inflammatory response
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
response to interleukin-1
3'-UTR-mediated mRNA stabilization
cellular response to mechanical stimulus
death receptor binding
protein binding
identical protein binding
protein self-association
TIR domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0