This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290725_m1
See other PRSS55 GE Assays ›
Gene Symbol:
PRSS55
Gene Name:
protease, serine 55
Gene Aliases:
CT153, T-SP1, TSP1, UNQ9391
Chromosome Location:
Chr.8: 10525331 - 10554166 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290725_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197020.1 NP_001183949.1 1 - 2 194 96
NM_198464.3 NP_940866.2 1 - 2 194 96
XM_005272372.3 XP_005272429.2 1 - 2 401 96
XM_011543814.2 XP_011542116.2 1 - 2 409 96
XM_017013182.1 XP_016868671.1 1 - 2 406 96
GenBank mRNA AY358867.1 - 1 - 2 169 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS55

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline