This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290861_m1
See other RIC3 GE Assays ›
Gene Symbol:
RIC3
Gene Name:
RIC3 acetylcholine receptor chaperone
Gene Aliases:
AYST720, PRO1385
Chromosome Location:
Chr.11: 8092965 - 8169088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290861_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206672.2 NP_001193601.1 3 - 4 484 114
GenBank mRNA AM422214.1 - 3 - 4 430 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of cytosolic calcium ion concentration
synaptic transmission, cholinergic
protein localization to cell surface
cellular protein complex assembly
acetylcholine receptor binding
protein binding involved in protein folding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0