This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291176_m1
See other SMUG1 GE Assays ›
Gene Symbol:
SMUG1
Gene Name:
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases:
FDG, HMUDG, UNG3
Chromosome Location:
Chr.12: 54171815 - 54188994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291176_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMUG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243788.1 NP_001230717.1 1 - 2 45 84
NM_001243791.1 NP_001230720.1 1 - 2 45 84
GenBank mRNA BG706635.1 - 1 - 2 48 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001243788.1, NM_001243791.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
depyrimidination
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
DNA binding
uracil DNA N-glycosylase activity
protein binding
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0