This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291182_m1
See other NUDT2 GE Assays ›
Gene Symbol:
NUDT2
Gene Name:
nudix hydrolase 2
Gene Aliases:
APAH1
Chromosome Location:
Chr.9: 34329506 - 34343713 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291182_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244390.1 NP_001231319.1 1 - 2 89 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001244390.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
nucleobase-containing compound metabolic process
apoptotic process
bis(5'-nucleosyl)-tetraphosphatase (asymmetrical) activity
protein binding
GTP binding
bis(5'-nucleosyl)-tetraphosphatase (symmetrical) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0