This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291384_m1
See other TMEM254 GE Assays ›
Gene Symbol:
TMEM254
Gene Name:
transmembrane protein 254
Gene Aliases:
C10orf57, bA369J21.6
Chromosome Location:
Chr.10: 80078646 - 80092552 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291384_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM254
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270369.1 NP_001257298.1 4 - 5 379 98
NM_001270373.1 NP_001257302.1 4 - 5 358 98
NR_072987.1 4 - 5 280 98
GenBank mRNA BP293793.1 - 4 - 5 333 98
DA263618.1 - 4 - 5 238 98
DA573956.1 - 4 - 5 339 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline