This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291550_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077702.2 NP_001071170.2 2 - 3 301 99
NM_001146111.1 NP_001139583.1 2 - 3 301 99
NM_001278215.1 NP_001265144.1 2 - 3 301 99
NM_020948.3 NP_065999.2 2 - 3 301 99
XM_005271076.3 XP_005271133.1 2 - 3 339 99
XM_011541864.2 XP_011540166.1 2 - 3 209 99
XM_017001919.1 XP_016857408.1 2 - 3 186 99
XM_017001920.1 XP_016857409.1 2 - 3 203 99
XM_017001923.1 XP_016857412.1 2 - 3 209 99
GenBank mRNA AF515448.2 - 2 - 3 238 99
AY124186.2 - 2 - 3 238 99
AY124189.2 - 2 - 3 238 99
AY124192.2 - 2 - 3 238 99
BC066898.1 - 2 - 3 229 99
BC108726.1 - 2 - 3 182 5194296 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF515448.2, AY124186.2, AY124189.2, AY124192.2, BC066898.1, BC108726.1, NM_001077702.2, NM_001146111.1, NM_001278215.1, NM_020948.3, XM_005271076.3, XM_017001919.1, XM_017001920.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MIER1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0