This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291613_m1
See other LIAS GE Assays ›
Gene Symbol:
LIAS
Gene Name:
lipoic acid synthetase
Gene Aliases:
HGCLAS, HUSSY-01, LAS, LIP1, LS, PDHLD
Chromosome Location:
Chr.4: 39459022 - 39477653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291613_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIAS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278590.1 NP_001265519.1 1 - 2 144 105
NM_001278591.1 NP_001265520.1 1 - 2 144 105
NM_001278592.1 NP_001265521.1 1 - 2 144 105
NM_006859.3 NP_006850.2 1 - 2 144 105
NM_194451.2 NP_919433.1 1 - 2 144 105
XM_006713990.3 XP_006714053.1 1 - 2 146 105
XM_017007665.1 XP_016863154.1 1 - 2 137 105
XM_017007666.1 XP_016863155.1 1 - 2 146 105
XM_017007667.1 XP_016863156.1 1 - 2 137 105
GenBank mRNA AK026289.1 - 1 - 2 112 105
AK291985.1 - 1 - 2 75 105
AK292238.1 - 1 - 2 144 105
AK303429.1 - 1 - 2 123 105
AV729111.1 - 1 - 2 129 105
BC023635.2 - 1 - 2 89 4841470 105
BC062751.1 - 1 - 2 114 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK291985.1, AK292238.1, AK303429.1, BC023635.2, BC062751.1, NM_001278590.1, NM_001278591.1, NM_001278592.1, NM_006859.3, NM_194451.2, XM_006713990.3, XM_017007665.1, XM_017007666.1, XM_017007667.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LIAS

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
inflammatory response
response to oxidative stress
lipoate biosynthetic process
protein lipoylation
response to lipopolysaccharide
glyoxylate metabolic process
lipoate synthase activity
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0