This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291614_m1
See other LIAS GE Assays ›
Gene Symbol:
LIAS
Gene Name:
lipoic acid synthetase
Gene Aliases:
HGCLAS, HUSSY-01, LAS, LIP1, LS, PDHLD
Chromosome Location:
Chr.4: 39459022 - 39477653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291614_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIAS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278592.1 NP_001265521.1 2 - 3 310 99
XM_006713990.3 XP_006714053.1 2 - 3 312 99
XM_017007666.1 XP_016863155.1 2 - 3 312 99
GenBank mRNA AI373464.1 - 1 - 2 301 99
AK303429.1 - 2 - 3 289 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
inflammatory response
response to oxidative stress
lipoate biosynthetic process
protein lipoylation
response to lipopolysaccharide
glyoxylate metabolic process
lipoate synthase activity
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0