This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291675_gH
See other EEF1D GE Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1D, EF1D, FP1047
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291675_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130053.2 NP_001123525.2 4 - 5 1556 105
NM_001130055.2 NP_001123527.1 3 - 4 627 105
NM_001130057.2 NP_001123529.1 2 - 3 633 105
NM_001195203.1 NP_001182132.1 2 - 3 451 105
NM_001289950.1 NP_001276879.1 2 - 3 448 105
NM_001960.4 NP_001951.2 2 - 3 451 105
NM_032378.4 NP_115754.3 4 - 5 1673 105
XM_005250820.3 XP_005250877.1 3 - 4 6548 105
XM_005250823.1 XP_005250880.1 3 - 4 1282 105
XM_006716519.3 XP_006716582.1 3 - 4 6545 105
XM_006716522.1 XP_006716585.1 3 - 4 1536 105
XM_006716523.1 XP_006716586.1 4 - 5 1496 105
XM_006716524.1 XP_006716587.1 3 - 4 1303 105
XM_006716525.1 XP_006716588.1 2 - 3 471 105
XM_011516905.2 XP_011515207.1 4 - 5 6118 105
XM_011516906.2 XP_011515208.1 6 - 7 4710 105
XM_017013168.1 XP_016868657.1 4 - 5 5549 105
XM_017013169.1 XP_016868658.1 5 - 6 5205 105
XM_017013170.1 XP_016868659.1 4 - 5 4991 105
XM_017013171.1 XP_016868660.1 3 - 4 1279 105
GenBank mRNA AF370363.1 - 4 - 5 1256 105
AK024550.1 - 4 - 5 1410 105
AK093756.1 - 2 - 3 1593 105
AK225040.1 - 4 - 5 1411 105
AK225243.1 - 2 - 3 188 105
AK225270.1 - 2 - 3 189 105
AK293339.1 - 2 - 3 189 105
AK314311.1 - 4 - 5 1292 105
BC007847.2 - 4 - 5 1277 4129368 105
BC009907.2 - 1 173 105
BC012819.2 - 1 173 105
BC062535.1 - 2 - 3 165 105
BC071840.1 - 2 - 3 153 105
BI667117.1 - 2 - 3 456 105
BQ064320.1 - 308 105
BQ686171.1 - 3 - 4 346 105
BT007242.1 - 2 - 3 1214 105
CN424083.1 - 2 - 3 392 105
CN424105.1 - 2 - 3 236 105
CX787126.1 - 2 - 3 498 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1D

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0