This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291678_gH
See other PRSS3 GE Assays ›
Gene Symbol:
PRSS3
Gene Name:
protease, serine 3
Gene Aliases:
MTG, PRSS4, T9, TRY3, TRY4
Chromosome Location:
Chr.9: 33750466 - 33799231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291678_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197097.2 NP_001184026.2 5 - 6 939 113
NM_001197098.1 NP_001184027.1 4 - 5 581 113
NM_002771.3 NP_002762.2 4 - 5 602 113
NM_007343.3 NP_031369.2 4 - 5 811 113
XM_011517965.1 XP_011516267.1 5 - 6 862 113
GenBank mRNA AB298285.1 - 4 - 5 811 113
AB298286.1 - 4 - 5 581 113
BC030238.1 - 4 - 5 601 113
BC069476.1 - 4 - 5 602 7262223 113
BC069494.1 - 4 - 5 602 7262235 113
D45417.1 - 4 - 5 589 113
X71345.1 - 4 - 5 629 113
X72781.1 - 4 - 5 632 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS3

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
cobalamin metabolic process
zymogen activation
endothelial cell migration
serine-type endopeptidase activity
calcium ion binding
protein binding
serine-type peptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline