This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291701_g1
See other UBE2F GE Assays ›
Gene Symbol:
UBE2F
Gene Name:
ubiquitin conjugating enzyme E2 F (putative)
Gene Aliases:
NCE2
Chromosome Location:
Chr.2: 237966945 - 238042782 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291701_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2F
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278305.1 NP_001265234.1 9 - 10 643 77
NM_001278306.1 NP_001265235.1 8 - 9 580 77
NM_001278307.1 NP_001265236.1 7 - 8 618 77
NM_080678.2 NP_542409.1 9 - 10 714 77
NR_103498.1 8 - 9 646 77
NR_103499.1 7 - 8 623 77
NR_103500.1 10 - 11 813 77
GenBank mRNA AF310723.1 - 8 - 9 510 77
AK294107.1 - 8 - 9 526 77
AK297502.1 - 7 - 8 549 77
AK303094.1 - 7 - 8 572 77
BC010549.1 - 9 - 10 601 4153270 77
BC039435.1 - 1 - 2 813 77
BM470609.1 - 444 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UBE2F

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

postreplication repair
protein neddylation
protein K63-linked ubiquitination
protein binding
ATP binding
ligase activity
NEDD8 transferase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0