This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291711_gH
See other EEF1D GE Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1D, EF1D, FP1047
Chromosome Location:
Chr.8: 143579697 - 143597675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291711_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130053.2 NP_001123525.2 6 - 7 1726 121
NM_001130055.2 NP_001123527.1 5 - 6 797 121
NM_001130056.2 NP_001123528.1 3 - 4 549 121
NM_001130057.2 NP_001123529.1 4 - 5 803 121
NM_001289950.1 NP_001276879.1 4 - 5 618 121
NM_001317743.1 NP_001304672.1 3 - 4 546 121
NM_001960.4 NP_001951.2 4 - 5 621 121
NM_032378.4 NP_115754.3 6 - 7 1843 121
XM_005250820.3 XP_005250877.1 5 - 6 6718 121
XM_005250823.1 XP_005250880.1 5 - 6 1452 121
XM_006716519.3 XP_006716582.1 5 - 6 6715 121
XM_006716520.3 XP_006716583.1 4 - 5 6646 121
XM_006716522.1 XP_006716585.1 5 - 6 1706 121
XM_006716523.1 XP_006716586.1 6 - 7 1666 121
XM_006716524.1 XP_006716587.1 5 - 6 1473 121
XM_006716525.1 XP_006716588.1 4 - 5 641 121
XM_011516905.2 XP_011515207.1 6 - 7 6288 121
XM_011516906.2 XP_011515208.1 8 - 9 4880 121
XM_017013168.1 XP_016868657.1 6 - 7 5719 121
XM_017013169.1 XP_016868658.1 7 - 8 5375 121
XM_017013170.1 XP_016868659.1 6 - 7 5161 121
XM_017013171.1 XP_016868660.1 5 - 6 1449 121
XM_017013172.1 XP_016868661.1 4 - 5 1634 121
XM_017013173.1 XP_016868662.1 3 - 4 726 121
GenBank mRNA AF370363.1 - 6 - 7 1426 121
AK024550.1 - 6 - 7 1580 121
AK093756.1 - 4 - 5 1763 121
AK225040.1 - 6 - 7 1581 121
AK225243.1 - 4 - 5 358 121
AK225270.1 - 4 - 5 359 121
AK314311.1 - 6 - 7 1462 121
BC000678.2 - 3 - 4 1093 121
BC007847.2 - 6 - 7 1447 4129368 121
BC009907.2 - 1 343 121
BC012819.2 - 1 343 121
BC062535.1 - 4 - 5 335 121
BC071840.1 - 4 - 5 323 121
BC094806.1 - 3 - 4 260 121
BG482145.1 - 3 - 4 253 121
BI667117.1 - 4 - 5 626 121
BQ064320.1 - 478 121
BQ686171.1 - 5 - 6 516 121
BT007242.1 - 4 - 5 1384 121
CN424105.1 - 4 - 5 406 121
Z21507.1 - 4 - 5 355 121

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1D

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
cellular response to ionizing radiation
cell-cell adhesion
DNA binding
translation elongation factor activity
signal transducer activity
protein binding
translation factor activity, RNA binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0