This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291723_g1
See other RPL26L1 GE Assays ›
Gene Symbol:
RPL26L1
Gene Name:
ribosomal protein L26 like 1
Gene Aliases:
RPL26P1
Chromosome Location:
Chr.5: 172953677 - 172969771 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291723_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL26L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317980.1 NP_001304909.1 2 - 3 273 98
NM_001317981.1 NP_001304910.1 2 - 3 247 98
NM_001317982.1 NP_001304911.1 2 - 3 229 98
NM_016093.3 NP_057177.1 2 - 3 219 98
XM_005265921.3 XP_005265978.1 2 - 3 270 98
XM_011534565.2 XP_011532867.1 2 - 3 834 98
XM_017009519.1 XP_016865008.1 2 - 3 261 98
XM_017009520.1 XP_016865009.1 2 - 3 192 98
GenBank mRNA AF083248.1 - 2 - 3 193 98
AK128872.1 - 1 187 98
BC017360.2 - 2 - 3 207 4551093 98
BC070192.1 - 2 - 3 192 6727802 98
BI838595.1 - 247 98
BQ278240.1 - 2 - 3 220 98
BU193755.1 - 2 - 3 253 98
BU569345.1 - 2 - 3 232 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL26L1

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
ribosomal large subunit biogenesis
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0