This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291742_gH
See other PRSS3 GE Assays ›
Gene Symbol:
PRSS3
Gene Name:
protease, serine 3
Gene Aliases:
MTG, PRSS4, T9, TRY3, TRY4
Chromosome Location:
Chr.9: 33750466 - 33799231 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291742_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197097.2 NP_001184026.2 4 - 5 800 87
NM_001197098.1 NP_001184027.1 3 - 4 442 87
NM_002771.3 NP_002762.2 3 - 4 463 87
NM_007343.3 NP_031369.2 3 - 4 672 87
XM_011517965.1 XP_011516267.1 4 - 5 723 87
GenBank mRNA AB298285.1 - 3 - 4 672 87
AB298286.1 - 3 - 4 442 87
BC030238.1 - 3 - 4 462 87
BC069476.1 - 3 - 4 463 7262223 87
BC069494.1 - 3 - 4 463 7262235 87
D45417.1 - 3 - 4 450 87
X15505.1 - 3 - 4 463 87
X71345.1 - 3 - 4 490 87
X72781.1 - 3 - 4 493 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS3

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
cobalamin metabolic process
zymogen activation
endothelial cell migration
serine-type endopeptidase activity
calcium ion binding
protein binding
serine-type peptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline