This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07292226_m1
See other ADORA3 GE Assays ›
Gene Symbol:
ADORA3
Gene Name:
adenosine A3 receptor
Gene Aliases:
A3AR
Chromosome Location:
Chr.1: 111499429 - 111504121 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07292226_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADORA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001302678.1 NP_001289607.1 1 - 2 1122 109
GenBank mRNA CD013875.1 - 1 - 2 413 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

adenosine receptor signaling pathway
G-protein coupled adenosine receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0