This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07292253_s1
See other RPL10 GE Assays ›
Gene Symbol:
RPL10
Gene Name:
ribosomal protein L10
Gene Aliases:
AUTSX5, DXS648, DXS648E, L10, NOV, QM
Chromosome Location:
Chr.X: 154398065 - 154402339 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07292253_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303624.1 NP_001290553.1 1 122 92
GenBank mRNA AF218023.1 - 1 91 92
AK026568.1 - 1 119 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC065487.1 - 1 324 Chr. X - 154396701 - 154398816 [-] on Build GRCh38 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF218023.1, NM_001303624.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline