This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07292900_g1
See other PPP1R14B GE Assays ›
Gene Symbol:
PPP1R14B
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14B
Gene Aliases:
PHI-1, PLCB3N, PNG, SOM172
Chromosome Location:
Chr.11: 64244479 - 64246941 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07292900_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138689.2 NP_619634.1 1 225 108
GenBank mRNA BC014522.2 - 1 77 3831227 108
BC094817.1 - 1 108 5466876 108
BM847221.1 - 1 225 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC014522.2, BC094817.1, NM_138689.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R14B

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
regulation of phosphorylation
negative regulation of catalytic activity
innate immune response
protein serine/threonine phosphatase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0